Anti-SCYLV Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Toimittaja: Bioss
BS-2121R-A750
BOSSBS-2121R-A750EA 890 EUR
BOSSBS-2121R-A750
Anti-SCYLV Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Vasta-aineet
Yellow leaf disease of sugarcane (Saccharum spp.) is an important and widely spread disease causing severe yield losses, ranging from 20 to 40 % in susceptible cultivars. It is an emerging viral disease whose causal agent is a Polerovirus, the Sugarcane yellow leaf virus (SCYLV) transmitted by aphids.

Type: Primary
Antigen: SCYLV
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Virus, Others
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR