Anti-AQP9 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Toimittaja: Bioss
BS-2060R-A680
BOSSBS-2060R-A680EA 899 EUR
BOSSBS-2060R-A680
Anti-AQP9 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Vasta-aineet
Forms a channel with a broad specificity. Mediates passage of a wide variety of non-charged solutes including carbamides, polyols, purines, and pyrimidines in a phloretin- and mercury-sensitive manner, whereas amino acids, cyclic sugars, Na(+), K(+), Cl(-), and deprotonated monocarboxylates are excluded. Also permeable to urea and glycerol.

Type: Primary
Antigen: AQP9
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-295/295
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Others
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR