Anti-LXN Rabbit Polyclonal Antibody (Cy3®)

Toimittaja: Bioss
BS-1971R-CY3
BOSSBS-1971R-CY3EA 900 EUR
BOSSBS-1971R-CY3
Anti-LXN Rabbit Polyclonal Antibody (Cy3®)
Vasta-aineet
Latexin, a protein possessing inhibitory activity against rat carboxypeptidase A1 (CPA1) and CPA2, is expressed in a neuronal subset in the cerebral cortex and cells in other neural and non-neural tissues of rat. Although latexin also inhibits mast-cell CPA (MCCPA), the expression of latexin in rat mast cells has not previously been confirmed.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LXN
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR