Anti-Reprimo Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Toimittaja: Bioss
BS-1885R-A647
BOSSBS-1885R-A647EA 901 EUR
BOSSBS-1885R-A647
Anti-Reprimo Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Vasta-aineet
May be involved in the regulation of p53-dependent G2 arrest of the cell cycle. Seems to induce cell cycle arrest by inhibiting CDK1 activity and nuclear translocation of the CDC2 cyclin B1 complex (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Reprimo
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR