Anti-NGB Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Toimittaja: Bioss
BS-1859R-FITC
BOSSBS-1859R-FITCEA 900 EUR
BOSSBS-1859R-FITC
Anti-NGB Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Vasta-aineet
Involved in oxygen transport in the brain. Hexacoordinate globin, displaying competitive binding of oxygen or the distal His residue to the iron atom. Not capable of penetrating cell membranes. The deoxygenated form exhibits nitrite reductase activity inhibiting cellular respiration via NO-binding to cytochrome c oxidase. Involved in neuroprotection during oxidative stress. May exert its anti-apoptotic activity by acting to reset the trigger level of mitochondrial cytochrome c release necessary to commit the cells to apoptosis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary