Anti-IGLL1 Rabbit Polyclonal Antibody (Cy3)

Toimittaja: Bioss
BS-15577R-CY3
BOSSBS-15577R-CY3EA 900 EUR
BOSSBS-15577R-CY3
Anti-IGLL1 Rabbit Polyclonal Antibody (Cy3)
Vasta-aineet
The preB cell receptor is found on the surface of proB and preB cells, where it is involved in transduction of signals for cellular proliferation, differentiation from the proB cell to the preB cell stage, allelic exclusion at the Ig heavy chain gene locus, and promotion of Ig light chain gene rearrangements. The preB cell receptor is composed of a membrane-bound Ig mu heavy chain in association with a heterodimeric surrogate light chain. This gene encodes one of the surrogate light chain subunits and is a member of the immunoglobulin gene superfamily. This gene does not undergoMore Product Information
Order Now

TEKNISET TIEDOT

Antigeenin nimi IGLL1
Environmentally Preferable
Antigeenin synonyymit AGM2|Immunoglobulin omega polypeptide|CD179b antigen|VPREB2|IGL1|Ig lambda-5|Immunoglobulin-related protein 14,1|IGO|Ig lambda 5|CD179 antigen-like family member B|IGVPB|Immunoglobulin lambda-like polypeptide 1|Lambda5|IGLJ14,1|Igll1|Pre B lymphocyte specic protein 2|IGLL1_HUMAN|Immunoglobulin related protein 14,1|IGLL|CD179B|Immunoglobulin lambda like polypeptide 1 precursor|Immunoglobulin omega polypeptide chain|IGL5|14,1
Vasta-aineen tyyppi Ensisijainen
Klonaalisuus Polyclonal
Konjugaatio Cy3®
Reaktiivisuus Mouse
Isäntä Rabbit
Isotyyppi IgG
Western blot Yes
ImmunoKemia Yes
ImmunoFluoresenssi Yes
Modification Unmodified
Epitooppi 151-213/213
Immunogeeni KLH conjugated synthetic peptide derived from human IGLL1
Lomake Liquid
Molekyylipaino 19 kDa
Puhdistus Purified by Protein A
Pitoisuus 1 µg/µl
Säilytyspuskuri Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Säilytyslämpötila Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Kuljetuslämpötila 4 °C
Testatut aplikaatiot ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR