Anti-CCL26 Rabbit Polyclonal Antibody (FITC)

Toimittaja: Bioss
BS-15513R-FITC
BOSSBS-15513R-FITCEA 883 EUR
BOSSBS-15513R-FITC
Anti-CCL26 Rabbit Polyclonal Antibody (FITC)
Vasta-aineet
Eotaxin 3 is a recently discovered CC chemokine. It binds to the CCR3 receptor and is highly chemotactic for eosinophils and basophils.

Type: Primary
Antigen: CCL26
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 21-94/94
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR