Anti-TEC Rabbit Polyclonal Antibody
Toimittaja: Bioss
BS-15505R
BOSSBS-15505REA 725 EUR
BOSSBS-15505R
Anti-TEC Rabbit Polyclonal Antibody
Vasta-aineet
TEC is a belongs to the Tec family of non-receptor protein-tyrosine kinases containing a pleckstrin homology domain. Tec family kinases are involved in the intracellular signaling mechanisms of cytokine receptors, lymphocyte surface antigens, heterotrimeric G-protein coupled receptors, and integrin molecules. They are also key players in the regulation of the immune functions. Tec kinase is an integral component of T cell signaling and has a distinct role in T cell activation. This gene may be associated with myelodysplastic syndrome.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)