Anti-CYP2S1 Rabbit Polyclonal Antibody (ALEXA FLUOR® 488)

Toimittaja: Bioss
BS-15499R-A488
BOSSBS-15499R-A488EA 893 EUR
BOSSBS-15499R-A488
Anti-CYP2S1 Rabbit Polyclonal Antibody (ALEXA FLUOR® 488)
Vasta-aineet
CYP2S1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyse many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localises to the endoplasmic reticulum. In rodents, the homologous protein has been shown to metabolise certain carcinogens; however, the specific function of the human protein has not been determined.

Type: Primary
Antigen: CYP2S1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 488
Public Immunogen Range: 21-120/504
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR