Anti-CD46/MCP Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Toimittaja: Bioss
BS-1529R-A555
BOSSBS-1529R-A555EA 893 EUR
BOSSBS-1529R-A555
Anti-CD46/MCP Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Vasta-aineet
Acts as a cofactor for complement factor I, a serine protease which protects autologous cells against complement-mediated injury by cleaving C3b and C4b deposited on host tissue. May be involved in the fusion of the spermatozoa with the oocyte during fertilisation. Also acts as a costimulatory factor for T-cells which induces the differentiation of CD4+ into T-regulatory 1 cells. T-regulatory 1 cells suppress immune responses by secreting interleukin-10, and therefore are thought to prevent autoimmunity. A number of viral and bacterial pathogens seem to exploit this property and directlyMore Product Information
Order Now

TEKNISET TIEDOT

Environmentally Preferable
Antigeenin nimi CD46/MCP
Antigeenin synonyymit Membrane cofactor protein|Trophoblast leukocyte common antigen|CD46|MIC10|MCP|TRA2.10|AHUS2|TLX
Vasta-aineen tyyppi Ensisijainen
Klonaalisuus Polyclonal
Konjugaatio Alexa Fluor® 555
Reaktiivisuus Human,Mouse
Isäntä Rabbit
Geeni ID 4179
Isotyyppi IgG
Western blot Yes
ImmunoKemia Yes
ImmunoFluoresenssi Yes
Virtaussytometria Yes
Modification Unmodified
Epitooppi 251-355/355
Immunogeeni KLH conjugated synthetic peptide derived from Mouse CD46
Lomake Liquid
Molekyylipaino 43 kDa
Puhdistus Purified by Protein A
Pitoisuus 1 µg/µl
Säilytyspuskuri Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Säilytyslämpötila Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Kuljetuslämpötila 4 °C
Testatut aplikaatiot ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR