Anti-C1orf149 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Toimittaja: Bioss
BS-15027R-A488
BOSSBS-15027R-A488EA 893 EUR
BOSSBS-15027R-A488
Anti-C1orf149 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Vasta-aineet
C1orf149

Type: Primary
Antigen: C1orf149
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 488
Public Immunogen Range: 31-100/191
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR