Anti-IRAK2 Rabbit Polyclonal Antibody (HRP (Horseradish PE (Phycoerythrin)rOxidase))

Toimittaja: Bioss
BS-1427R-HRP
BOSSBS-1427R-HRPEA 900 EUR
BOSSBS-1427R-HRP
Anti-IRAK2 Rabbit Polyclonal Antibody (HRP (Horseradish PE (Phycoerythrin)rOxidase))
Vasta-aineet
IRAK or Interleukin-1 Receptor-associated Kinase 1, is one of two putative serine/threonine kinases that become associated with the interleukin-1 receptor (IL1R) upon stimulation. This protein is partially responsible for IL1-induced upregulation of the ubiquitous transcription factor NF-kappa B.

Type: Primary
Antigen: IRAK2
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public Immunogen Range: 21-150/625
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR