Anti-GPR44 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Toimittaja: Bioss
BS-13537R-A750
BOSSBS-13537R-A750EA 890 EUR
BOSSBS-13537R-A750
Anti-GPR44 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Vasta-aineet
Receptor for prostaglandin D2 (PGD2). Coupled to the G(i)-protein. Receptor activation may result in pertussis toxin-sensitive decreases in cAMP levels and Ca(2+) mobilization. PI3K signaling is also implicated in mediating PTGDR2 effects. PGD2 induced receptor internalization. CRTH2 internalization can be regulated by diverse kinases such as, PKC, PKA, ADRBK1/GRK2, GPRK5/GRK5 and GRK6. Receptor activation is responsible, at least in part, in immune regulation and allergic/inflammation responses.

Type: Primary
Antigen: GPR44
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
PublicMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR