Anti-GGTLC2 Rabbit Polyclonal Antibody (Cy7®)

Toimittaja: Bioss
BS-13353R-CY7
BOSSBS-13353R-CY7EA 900 EUR
BOSSBS-13353R-CY7
Anti-GGTLC2 Rabbit Polyclonal Antibody (Cy7®)
Vasta-aineet
GGTLC2

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GGTLC2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR