Anti-GGA1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Toimittaja: Bioss
BS-13343R-HRP
BOSSBS-13343R-HRPEA 900 EUR
BOSSBS-13343R-HRP
Anti-GGA1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Vasta-aineet
The GGA family of proteins (Golgi-localized, ARF-binding proteins) are ubiquitous coat proteins that facilitate the trafficking of soluble proteins from the trans-Golgi network (TGN) to endosomes/lysosomes by means of interactions with TGN-sorting receptors, ARF (ADP-ribosylation factor), and clathrin (1?). Members of the GGA family, GGA1,GGA2 (also known as VEAR) and GGA3, are multidomain proteins that bind mannose 6-phosphate receptors (MPRs) (1,2,4). GGAs have modular structures with an N-terminal VHS (VPS-27, Hrs, and STAM) domain followed by a GAT (GGA and TOM1) domain, a connectingMore Product Information
Order Now
TEKNISET TIEDOT
Antigeenin symboli GGA1
Environmentally Preferable
Antigeenin nimi Golgi-associated, Gamma Adaptin Ear Containing, ARF binding Protein 1
Antigeenin synonyymit gamma ear-containing|ADP-ribosylation factor binding protein GGA1|Gamma-adaptin related protein 1|ARF-binding protein 1|Golgi-localized
Vasta-aineen tyyppi Ensisijainen
Klonaalisuus Polyclonal
Konjugaatio HRP (Horseradish Peroxidase)
Reaktiivisuus Human,Rat,Mouse
Isäntä Rabbit
Geeni ID 26088
Isotyyppi IgG
Western blot Yes
ImmunoKemia Yes
Lomake liquid
Puhdistus Purified by Protein A
Pitoisuus 1 μg/μl
Ristiadsorptio No
Säilytyspuskuri Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
Säilytyslämpötila Store at 4°C for 12 months
Kuljetuslämpötila 4°C