Anti-GADL1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Toimittaja: Bioss
BS-13264R-A680
BOSSBS-13264R-A680EA 890 EUR
BOSSBS-13264R-A680
Anti-GADL1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Vasta-aineet
There are two forms of glutamic acid decarboxylases (GADs) that exist in brain: GAD-65 (also known as GAD2) and GAD-67 (also known as GAD1, GAD or SCP). GAD-65 and GAD-67 are members of the group II decarboxylase family of proteins and are responsible for catalyzing the rate limiting step in the production of GABA (gamma-aminobutyric acid) from L-glutamic acid. Although both GADs are found in brain, GAD-65 localizes to synaptic vesicle membranes in nerve terminals, while GAD-67 is distributed throughout the cell. GAD-67 is responsible for the basal levels of GABA synthesis. In the case of aMore Product Information