Anti-FDPS Rabbit Polyclonal Antibody (Cy7®)
Toimittaja: Bioss
BS-13153R-CY7
BOSSBS-13153R-CY7EA 893 EUR
BOSSBS-13153R-CY7
Anti-FDPS Rabbit Polyclonal Antibody (Cy7®)
Vasta-aineet
FDPS is a 419 amino acid enzyme belonging to the FPP/GGPP synthetase family. Localized to cytoplasm and peroxisome, FDPS expression is regulated by phorbol esters and polyunsaturated fatty acids. FDPS assists in cholesterol biosynthesis, post-translational protein modifications and synthesis of steroid hormones in the isoprenoid pathway.FDPS catalyzes the formation of farnesyl diphosphate (FPP), a precursor for several classes of essential metabolites including sterols, dolichols, carotenoids, and ubiquinones. FDPS is inactivated by interferon-induced RSAD2, which may result in theMore Product Information