Anti-CAP1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Toimittaja: Bioss
BS-1306R-A488
BOSSBS-1306R-A488EA 893 EUR
BOSSBS-1306R-A488
Anti-CAP1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Vasta-aineet
Directly regulates filament dynamics and has been implicated in a number of complex developmental and morphological processes, including mRNA localization and the establishment of cell polarity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CAP1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR