Anti-TRDMT1 Rabbit Polyclonal Antibody
Toimittaja: Bioss
BS-13025R
BOSSBS-13025REA 725 EUR
BOSSBS-13025R
Anti-TRDMT1 Rabbit Polyclonal Antibody
Vasta-aineet
Methylation at the 5'-position of cytosine is the only known naturally occurring covalent modification of the mammalian genome. DNA methylation requires the enzymatic activity of DNA 5-cytosine methyltransferase (Dnmt) proteins, which catalyze the transfer of a methyl group from S-adenosyl methionine to the 5'-position of cytosines residing in the dinucleotide CpG motif, and this methylation results in transcriptional repression of the target gene. The Dnmt enzymes are encoded by independent genes. Dnmt1 is the most abundant, and it preferentially methylates hemimethylated DNA andMore Product Information