Anti-DIAPH2/Dia2 Rabbit Polyclonal Antibody

Toimittaja: Bioss
BS-13002R
BOSSBS-13002REA 725 EUR
BOSSBS-13002R
Anti-DIAPH2/Dia2 Rabbit Polyclonal Antibody
Vasta-aineet
Could be involved in oogenesis. Involved in the regulation of endosome dynamics. Implicated in a novel signal transduction pathway, in which isoform 3 and CSK are sequentially activated by RHOD to regulate the motility of early endosomes through interactions with the actin cytoskeleton.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: DIAPH2/Dia2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR