Anti-Blood Group Lewis a Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Toimittaja: Bioss
BS-12870R-A680
BOSSBS-12870R-A680EA 890 EUR
BOSSBS-12870R-A680
Anti-Blood Group Lewis a Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Vasta-aineet
Glycosyltransferases that mediate the region and stereoselective transfer of sugars, such as the fucosyltransferases, determine cell surface-carbohydrate profiles, which is an essential interface for biological recognition processes. Fucosyltransferases catalyse the covalent association of fucose to different positional linkages in sugar acceptor molecules. The carbohydrate moieties generated and covalently attached to cell surfaces are necessary to ensure a surface contour that satisfies physiological roles, which are reliant on adhesion molecules such as Selectins. Hematopoietic lineagesMore Product Information