Anti-CD40L Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Toimittaja: Bioss
BS-1286R-FITC
BOSSBS-1286R-FITCEA 909 EUR
BOSSBS-1286R-FITC
Anti-CD40L Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Vasta-aineet
Mediates B-cell proliferation in the absence of co-stimulus as well as IgE production in the presence of IL-4. Involved in immunoglobulin class switching. Release of soluble CD4L from platelets is partially regulated by GP IIb/IIIa, actin polymerization, and an matrix metalloproteinases (MMP) inhibitor-sensitive pathway.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD40L
Clonality: Polyclonal
Clone: 0
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR