Anti-ATP6V1B2 Rabbit Polyclonal Antibody
Toimittaja: Bioss
BS-12549R
BOSSBS-12549REA 725 EUR
BOSSBS-12549R
Anti-ATP6V1B2 Rabbit Polyclonal Antibody
Vasta-aineet
Vacuolar-type H+-ATPase (V-ATPase) is a multisubunit enzyme responsible for acidification of eukaryotic intracellular organelles. V-ATPases pump protons against an electrochemical gradient, while F-ATPases reverse the process, thereby synthesizing ATP. A peripheral V1 domain, which is responsible for ATP hydrolysis, and a integral V0 domain, which is responsible for proton translocation, compose V-ATPase. Nine subunits (A–H) make up the V1 domain and five subunits (a, d, c, c' and c") make up the V0 domain. Like F-ATPase, V-ATPase most likely operates through a rotary mechanism. The V-More Product Information