Anti-SPAG9 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Toimittaja: Bioss
BS-12270R-HRP
BOSSBS-12270R-HRPEA 900 EUR
BOSSBS-12270R-HRP
Anti-SPAG9 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Vasta-aineet
JIP-4 is a 1,321 amino acid protein encoded by the human gene SPAG9. It contains a large N-terminal extracellular domain, a short transmembrane helical domain, and a cytoplasmic domain. There are 6 N-glycosylation sites, several phosphorylation sites for cAMP/cGMP-dependent protein kinase, protein kinase C, and casein kinase II, and 10 putative myristoylation sites. There is also a leucine zipper motif, with 6 leucine repeats, that may aid in dimerization since there is no upstream basic domain characteristic of DNA binding proteins. The JNK-interacting protein (JIP) group of scaffoldMore Product Information