Anti-DAZL Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Toimittaja: Bioss
BS-12245R-A680
BOSSBS-12245R-A680EA 874 EUR
BOSSBS-12245R-A680
Anti-DAZL Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Vasta-aineet
The DAZ (Deleted in AZoospermia) gene family encodes potential RNA binding proteins that are expressed in prenatal and postnatal germ cells of males and females. The protein encoded by this gene is localised to the nucleus and cytoplasm of fetal germ cells and to the cytoplasm of developing oocytes. In the testis, this protein is localised to the nucleus of spermatogonia but relocates to the cytoplasm during meiosis where it persists in spermatids and spermatozoa. Transposition and amplification of this autosomal gene during primate evolution gave rise to the DAZ gene cluster on the YMore Product Information