Anti-MIR16/GDE1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Toimittaja: Bioss
BS-12039R-A750
BOSSBS-12039R-A750EA 890 EUR
BOSSBS-12039R-A750
Anti-MIR16/GDE1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Vasta-aineet
GDE1 is a 331 amino acid multi-pass membrane protein that localizes to both the membrane and the cytoplasm and contains one GDPD domain. Expressed in a wide variety of tissues, GDE1 uses magnesium as a cofactor to catalyse the conversion of 1-(sn-glycero-3-phospho)-1D-myo-inositol to myo-inositol and sn-glycerol 3-phosphate, an event that is modulated by G protein Signalling pathways and provides a link between phosphoinositide metabolism and G protein signal transduction. The gene encoding GDE1 maps to human chromosome 16, which encodes over 900 genes and comprises nearly 3% of the humanMore Product Information