Anti-CRBN Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Toimittaja: Bioss
BS-11716R-A750
BOSSBS-11716R-A750EA 890 EUR
BOSSBS-11716R-A750
Anti-CRBN Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Vasta-aineet
CRBN is a 442 amino acid protein which is highly concentrated in human brain tissue. CRBN functions are thought to be related to energy metabolism, learning and memory. localised to the cytoplasm, CRBN acts as a protease in mitochondria and is thought to regulate the assembly of KCNT1, as well as the surface expression of KCNT1 in brain regions known to affect memory and learning, such as the hippocampus. The gene encoding CRBN belongs to a family of ATP-dependent lon proteases that play a role in membrane trafficking and proteolysis. Defects in the CRBN gene are associated with mild mentalMore Product Information