Anti-GUCA2A Rabbit Polyclonal Antibody (Cy7®)
Toimittaja: Bioss
BS-11621R-CY7
BOSSBS-11621R-CY7EA 900 EUR
BOSSBS-11621R-CY7
Anti-GUCA2A Rabbit Polyclonal Antibody (Cy7®)
Vasta-aineet
The family of guanylin regulatory peptides, including guanylin and uroguanylin, are strongly expressed in intestinal mucosa and regulate intestinal fluid secretion during digestion. Guanylins are also involved in acid neutralization and the regulation of membrane-bound guanylate cyclase signaling molecules. Guanylin and uroguanylin are secreted primarily in the stomach, intestine, and colon. Guanylin is also detected in plasma. Guanylin is an endogenous activator of intestinal guanylate cyclase. It stimulates intestinal guanylate cyclase through the same receptor binding region as the heat-More Product Information