Anti-ZIC2 Rabbit Polyclonal Antibody
Toimittaja: Bioss
BS-11610R
BOSSBS-11610REA 725 EUR
BOSSBS-11610R
Anti-ZIC2 Rabbit Polyclonal Antibody
Vasta-aineet
Zic2 is a C2H2 zinc finger transcription factor that influences forebrain development. Zic2 is a transcriptional repressor and may regulate tissue specific expression of dopamine receptor D1. Zic2 transcript is abundant in the dorsal neural tube/spinal cord, and in the hindbrain. A polyhistidine tract polymorphism in this gene may be associated with increased risk of neural tube defects. This gene is closely linked to a gene encoding zinc finger protein of the cerebellum 5, a related family member on chromosome 13.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary