Anti-TBR1 Rabbit Polyclonal Antibody
Toimittaja: Bioss
BS-11603R
BOSSBS-11603REA 725 EUR
BOSSBS-11603R
Anti-TBR1 Rabbit Polyclonal Antibody
Vasta-aineet
A novel murine and human gene, TBR-1, encodes a putative transcription factor related to the Brachyrury (T) gene that is expressed only in postmitotic cells. T-brain-1 (TBR-1) mRNA is largely restricted to the cerebral cortex, where, during embryogenesis, it defines different regions that give rise to the palecortex, limbic cortex and neocortex (1-3). TBR-1, Pax-6 and Emx-1 are expressed in the mouse and chicken pallium. The pallio-subpallial boundary lies at the interface between the TBR-1 and Dlx-2 expression domains. Chicken genes homolgous to these mouse genes are expressed inMore Product Information