Anti-NOMO1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Toimittaja: Bioss
BS-11595R-A680
BOSSBS-11595R-A680EA 890 EUR
BOSSBS-11595R-A680
Anti-NOMO1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Vasta-aineet
Three highly similar proteins termed NOMO1, NOMO2 and NOMO3, are encoded by a gene mapping to a region of duplication on the p arm of human chromosome 16. All three NOMO proteins share similar functions and have been difficult to characterize individually. NOMO1 (Nodal modulator 1), also known as PM5, is a 1222 amino acid highly conserved single-pass type I membrane protein expressed in colon tumour tissue and normal colonic mucosa. NOMO proteins are novel antagonists of Nodal Signalling which interact with Nicalin to form a Nicalin-NOMO complex, and are rapidly degraded or stabilized byMore Product Information