Anti-SRC Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Toimittaja: Bioss
BS-1135R-A750
BOSSBS-1135R-A750EA 890 EUR
BOSSBS-1135R-A750
Anti-SRC Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Vasta-aineet
c-Src tyrosine kinase plays a critical role in signal transduction downstream of growth factor receptors, integrins and G protein-coupled receptors. We used stable isotope labelling with amino acids in cell culture (SILAC) approach to identify additional substrates of c-Src tyrosine kinase in human embryonic kidney 293T cells. We have identified 10 known substrates and interactors of c-Src and Src family kinases along with 26 novel substrates. We have experimentally validated 4 of the novel proteins (NICE-4, RNA binding motif 10, FUSE-binding protein 1 and TRK-fused gene) as directMore Product Information