Anti-Bestrophin 2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Toimittaja: Bioss
BS-11041R-A750
BOSSBS-11041R-A750EA 890 EUR
BOSSBS-11041R-A750
Anti-Bestrophin 2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Vasta-aineet
The Bestrophins are a newly described family of anion channels unrelated in primary sequence to any previously characterised channel proteins. Bestrophins were originally defined as a family of over 20 related sequences of the C. elegans. The first mammalian Bestrophin was identified as the vitelliform macular dystrophy (VMD), 1 also known as Best disease. Three more members of the bestrophin family members were cloned and indentified recently, Bestrophin 2, 3 and 4. RT PCR analyses revealed tissue restricted expression of the three genes with both Bestrophin 1 and Bestrophin 2 areMore Product Information