Anti-CCL26 Signal peptide Rabbit Polyclonal Antibody

Toimittaja: Bioss
BS-10409R
BOSSBS-10409REA 725 EUR
BOSSBS-10409R
Anti-CCL26 Signal peptide Rabbit Polyclonal Antibody
Vasta-aineet
Eotaxin 3 is a recently discovered CC chemokine. It binds to the CCR3 receptor and is highly chemotactic for eosinophils and basophils.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: CCL26 Signal peptide
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR