Anti-VGF/NERP2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Toimittaja: Bioss
BS-10332R-A750
BOSSBS-10332R-A750EA 890 EUR
BOSSBS-10332R-A750
Anti-VGF/NERP2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Vasta-aineet
This gene is specifically expressed in a subpopulation of neuroendocrine cells, and is upregulated by nerve growth factor. The structural organization of this gene is similar to that of the rat gene, and both the translated and the untranslated regions show a high degree of sequence similarity to the rat gene. The encoded secretory protein also shares similarities with the secretogranin/chromogranin family, however, its exact function is not known. Nerve growth factor (NGF) is a peptide that plays a key role in the differentiation and survival of neurons in the peripheral nervous system (PNS)More Product Information