Anti-CD38 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Toimittaja: Bioss
BS-0980R-A680
BOSSBS-0980R-A680EA 890 EUR
BOSSBS-0980R-A680
Anti-CD38 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Vasta-aineet
synthesises the second messagers cyclic ADP-ribose and nicotinate-adenine dinucleotide phosphate, the former a second messenger for glucose-induced insulin secretion. Also has cADPr hydrolase activity. Also moonlights as a receptor in cells of the immune system.
Type: Primary
Antigen: CD38
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 81-180/300
Host: Rabbit
Isotype: IgG
Reactivity: Human