Anti-Measles virus fusion Protein Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Toimittaja: Bioss
BS-0886R-A488
BOSSBS-0886R-A488EA 877 EUR
BOSSBS-0886R-A488
Anti-Measles virus fusion Protein Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Vasta-aineet
Class I viral fusion protein. Under the current model, the protein has at least 3 conformational states: pre-fusion native state, pre-hairpin intermediate state, and post-fusion hairpin state. During viral and plasma cell membrane fusion, the heptad repeat (HR) regions assume a trimer-of-hairpins structure, positioning the fusion peptide in close proximity to the C-terminal region of the ectodomain. The formation of this structure appears to drive apposition and subsequent fusion of viral and plasma cell membranes. Directs fusion of viral and cellular membranes leading to delivery of theMore Product Information