Anti-DMT1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Toimittaja: Bioss
BS-0653R-HRP
BOSSBS-0653R-HRPEA 900 EUR
BOSSBS-0653R-HRP
Anti-DMT1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Vasta-aineet
Important in metal transport, in particular iron. Can also transport manganese, cobalt, cadmium, nickel, vanadium and lead. Involved in apical iron uptake into duodenal enterocytes.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: Dmt1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR