Anti-ITGA5 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Toimittaja: Bioss
BS-0567R-A350
BOSSBS-0567R-A350EA 901 EUR
BOSSBS-0567R-A350
Anti-ITGA5 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Vasta-aineet
Integrin alpha-5/beta-1 is a receptor for fibronectin and fibrinogen. It recognizes the sequence R-G-D in its ligands. In case of HIV-1 infection, the interaction with extracellular viral Tat protein seems to enhance angiogenesis in Kaposi's sarcoma lesions.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ITGA5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR