Anti-MMP1 Rabbit Polyclonal Antibody (Cy7®)

Toimittaja: Bioss
BS-0424R-CY7
BOSSBS-0424R-CY7EA 901 EUR
BOSSBS-0424R-CY7
Anti-MMP1 Rabbit Polyclonal Antibody (Cy7®)
Vasta-aineet
Cleaves collagens of types I, II, and III at one site in the helical domain. Also cleaves collagens of types VII and X. In case of HIV infection, interacts and cleaves the secreted viral Tat protein, leading to a decrease in neuronal Tat's mediated neurotoxicity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MMP1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR