Anti-SLC11A2 Rabbit Polyclonal Antibody (Cy7®)

Toimittaja: Bioss
BS-0401R-CY7
BOSSBS-0401R-CY7EA 900 EUR
BOSSBS-0401R-CY7
Anti-SLC11A2 Rabbit Polyclonal Antibody (Cy7®)
Vasta-aineet
No data available.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC11A2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR