Anti-AK1 Rabbit Polyclonal Antibody (Cy5.5®)
Toimittaja: Bioss
BS-0299R-CY5.5
BOSSBS-0299R-CY5.5EA 900 EUR
BOSSBS-0299R-CY5.5
Anti-AK1 Rabbit Polyclonal Antibody (Cy5.5®)
Vasta-aineet
Adenylate kinase is an enzyme involved in regulating the adenine nucleotide composition within a cell by catalyzing the reversible transfer of phosphate group among adinine nucleotides. Three isozymes of adenylate kinase have been identified in vertebrates, adenylate isozyme 1 (AK1), 2 (AK2) and 3 (AK3). AK1 is found in the cytosol of skeletal muscle, brain and erythrocytes, whereas AK2 and AK3 are found in the mitochondria of other tissues including liver and heart. AK1 was identified because of its association with a rare genetic disorder causing nonspherocytic hemolytic anemia where aMore Product Information