Anti-TRKA Rabbit Polyclonal Antibody (Cy7®)
Toimittaja: Bioss
BS-0193R-CY7
BOSSBS-0193R-CY7EA 909 EUR
BOSSBS-0193R-CY7
Anti-TRKA Rabbit Polyclonal Antibody (Cy7®)
Vasta-aineet
Receptor tyrosine kinase involved in the development and the maturation of the central and peripheral nervous systems through regulation of proliferation, differentiation and survival of sympathetic and nervous neurons. High affinity receptor for NGF which is its primary ligand, it can also bind and be activated by NTF3/neurotrophin-3. However, NTF3 only supports axonal extension through NTRK1 but has no effect on neuron survival. Upon dimeric NGF ligand-binding, undergoes homodimerization, autophosphorylation and activation. Recruits, phosphorylates and/or activates several downstreamMore Product Information