Anti-SP Rabbit Polyclonal Antibody (Cy5.5®)

Toimittaja: Bioss
BS-0065R-CY5.5
BOSSBS-0065R-CY5.5EA 909 EUR
BOSSBS-0065R-CY5.5
Anti-SP Rabbit Polyclonal Antibody (Cy5.5®)
Vasta-aineet
Tachykinins are active peptides which excite neurons, evoke behavioral responses, are potent vasodilators and secretagogues, and contract (directly or indirectly) many smooth muscles.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SP
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR