Safety spectacles, Slam
Toimittaja: Bollé Safety
SLAPSI SLAESP SLAWPCC5
111-1014EA 8.7 EUR
111-1014 BOLESLAESP BOLESLAWPCC5
Safety spectacles, Slam
Suojalasit
Modern design spectacles with extra-easy fitting and comfort.
  • Panoramic visual field
  • Anti-scratch, anti-fog, anti-static coatings
  • Ultra enveloping
  • Comfort temples
Weight: 23 g.
Sertifikaatit: EN 166, EN 170 (clear lens), EN 172 (smoke lens).
Toimitustiedot: Supplied with adjustable cord.