Tubings, PFA, flexible, BOLA
Toimittaja: Bohlender
S1822-24 S1822-02 S1822-04 S1822-20 S1822-22 S1822-06 S1822-26 S1822-36 S1822-08 S1822-56 S1822-01 S1822-19 S1822-34 S1822-64 S1822-72 S1822-98 S1822-94 S1822-93 S1822-92 S1822-84 S1822-76 S1822-68 S1822-60 S1822-52 S1822-32 S1822-28 S1822-18 S1822-16 S1822-14 S1822-10 S1822-80
BOHLS1822-24EA 158 EUR
BOHLS1822-24 BOHLS1822-02 BOHLS1822-04 BOHLS1822-20 BOHLS1822-22 BOHLS1822-06 BOHLS1822-26 BOHLS1822-36 BOHLS1822-08 BOHLS1822-56 BOHLS1822-01 BOHLS1822-19 BOHLS1822-34 BOHLS1822-64 BOHLS1822-72 BOHLS1822-98 BOHLS1822-94 BOHLS1822-93 BOHLS1822-92 BOHLS1822-84 BOHLS1822-76 BOHLS1822-68 BOHLS1822-60 BOHLS1822-52 BOHLS1822-32 BOHLS1822-28 BOHLS1822-18 BOHLS1822-16 BOHLS1822-14 BOHLS1822-10 BOHLS1822-80
Tubings, PFA, flexible, BOLA
Letku Siirtoputki
PFA, translucent.
  • Non-porous and translucent, FDA conform
  • Temperature resistance: from −270 to +260 °C
  • Usable with a small bending radius
Flexible to highly flexible corrugated tubing with circular corrugations around the longitudinal axis. Cylindrical tubings ends with a length of 40 mm can for example be connected directly to fittings, stopcocks or hose connectors using BOLA Laboratory Screw Joints to provide absolutely tight connections which are also suitable for vacuum. Tight bending radius only causes little cross-section reduction. Ideal forMore Product Information