Bore adapter, b.safe
Toimittaja: Bohlender
R1169-54
BOHLR1169-54EA 119 EUR
BOHLR1169-54
Bore adapter, b.safe
Sovittimet Sovittimet, NS-hios
Made of PE, to reduce the bore diameter in table- or countertops from approx. 54 to 30 mm to ensure a safe installation of e.g. b.safe Desktop Mounts or b.safe Multiports in corresponding bores. Consisting of top part, lock washer and silicone gasket to avoid rotating and slipping during installation.
  • Ensure a safe installation
  • Avoid rotating and slipping during installation
  • Adaptation of existing bores
Adaptation of existing bores for assembling b.safe Desktop Mounts or b.safe Multiports also in bores with diameter bigger than 30 mm. EasyMore Product Information