Screw caps for bottles with GL threads, BOLA
Toimittaja: Bohlender
H993-18 H993-14 H993-25 H993-32 H993-45
BOHLH993-18EA 3.7 EUR
BOHLH993-18 BOHLH993-14 BOHLH993-25 BOHLH993-32 BOHLH993-45
Screw caps for bottles with GL threads, BOLA
Tulpat ja korkit Pullojen sulkimet
Screw cap made of PPS, with handy knurl and integrated PTFE/silicone gasket.
  • Very good chemical resistance
  • Temperature resistant from −20 to +230 °C
Suitable for bottles and vessels with GL threads.