Threaded couplings
Toimittaja: Bohlender
H901-01 H901-05 H900-02 H900-03 H901-02 H901-03 H900-01 H900-05 H901-04 H900-04
BOHLH901-01EA 37.2 EUR
BOHLH901-01 BOHLH901-05 BOHLH900-02 BOHLH900-03 BOHLH901-02 BOHLH901-03 BOHLH900-01 BOHLH900-05 BOHLH901-04 BOHLH900-04
Threaded couplings
Kiinnitykset ja liittimet
Connector, screw thread coupling, made of PTFE. Two screw caps made of PPS or PBTP with GL thread and a PTFE/FPM gasket. The product is only exposed to PTFE.
  • Applicable for connecting two GL threads of different sizes
  • Temperature resisitance from −200 to +250 ºC
  • Very good chemical resistance
  • FDA conformity