Screws with countersunk, BOLA
Toimittaja: Bohlender
H1124-26 H1124-22 H1124-18 H1124-14
BOHLH1124-26EA 26.3 EUR
BOHLH1124-26 BOHLH1124-22 BOHLH1124-18 BOHLH1124-14
Screws with countersunk, BOLA
Kiinnikkeet Screws
PTFE, white.
  • Temperature resistance: from -200 °C to +250 °C
  • Universal chemical resistance
  • FDA conform
  • Similar to DIN 963/DIN EN ISO 2009